Influenza A H6N4 (A/chicken/Hong Kong/17/1977) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40027-V08H-100
103663-528EA 351.48 USD
103663-528
Influenza A H6N4 (A/chicken/Hong Kong/17/1977) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain of influenza A virus hemagglutinin (A/chicken/Hong Kong/17/1977 (H6N4)) (CAC84244.1) (Met 1-Gln 529), was fused with a polyhistidine tag at the C-terminus.