Human Recombinant PCSK9/NARC1 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
29698-H05H-50 29698-H05H-50
103918-878EA 300.98 USD
103918-878 CA76488-010
Human Recombinant PCSK9/NARC1 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human PCSK9 ( NP_777596.2) (Met1-Gln692) was expressed with the Fc region of mouse IgG1 at the C-terminus.