Influenza A H15N8 Recombinant (A/duck/AUS/341/1983) Hemagglutinin/HA (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
11720-V08H-100
103622-726EA 351.48 USD
103622-726
Influenza A H15N8 Recombinant (A/duck/AUS/341/1983) Hemagglutinin/HA (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Val 534) of the influenza A H15N8 hemagglutinin (A/duck/AUS/341/1983 (H15N8)) (ABB88132.1) (HA1+HA2, uncleaved) was expressed, fused with a C-terminal polyhistidine tag.