Human Recombinant CD38 Protein (ECD, Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
10818-H02H-50 10818-H02H-50
103885-862EA 179.78 USD
CA76475-968 103885-862
Human Recombinant CD38 Protein (ECD, Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human CD38 (NP_001766.2) (Val43-Ile300) was expressed with the Fc region of human IgG1 at the C-terminus.