3-Amino-5-morpholinomethyl-2-oxazolidinone (AMOZ)-HRP Conjugate (1 mg)
Supplier: AAT Bioquest
50015
76484-044EA 580.89 USD
76484-044
3-Amino-5-morpholinomethyl-2-oxazolidinone (AMOZ)-HRP Conjugate (1 mg)
Proteins and Peptides
3-Amino-5-morpholinomethyl-2-oxazolidinone (AMOZ)-HRP conjugate is used for developing an ELISA assay for quantifying 3-Amino-5-morpholinomethyl-2-oxazolidinone (AMOZ).
  • Assay development
  • High sensitivity
Caution: For research use only.