Chloramphenicol-BSA Conjugate (1 mg)
Supplier: AAT Bioquest
50001
76484-016EA 580.89 USD
76484-016
Chloramphenicol-BSA Conjugate (1 mg)
Proteins and Peptides
Chloramphenicol-BSA conjugate is used for developing anti-chloramphenicol antibodies that are typically used in an ELISA assay.
  • Antibody development
  • High immunogenicity
Caution: For research use only.