iFluor® 488-Wheat Germ Agglutinin (WGA) Conjugate (1 mg)
Supplier: AAT Bioquest
25530
76485-356EA 107.15 USD
76485-356
iFluor® 488-Wheat Germ Agglutinin (WGA) Conjugate (1 mg)
Proteins and Peptides
Wheat germ agglutinin (WGA) is a lectin that binds to N-acetyl-D-glucosamine and sialic acid.
  • Glycoconjugate binding
  • Very bright signal
  • Multiple applications
Caution: For research use only.