Phalloidin-California Red Conjugate (300 Tests)
Supplier: AAT Bioquest
23103
76483-866EA 173.46 USD
76483-866
Phalloidin-California Red Conjugate (300 Tests)
Proteins and Peptides
This red fluorescent phalloidin conjugate (equivalent to Texas Red®-labeled phalloindin) selectively binds to F-actins with much higher photostability than the fluorescein-phalloidin conjugates.
  • Suitable for all fluorescent imaging platforms
  • Specific for actin in both living or fixed cells
  • Multiple applications
Caution: For research use only.