Phalloidin-AMCA Conjugate (300 Tests)
Supplier: AAT Bioquest
23100
76483-860EA 173.46 USD
76483-860
Phalloidin-AMCA Conjugate (300 Tests)
Proteins and Peptides
This blue fluorescent phalloidin conjugate (equivalent to coumarin-labeled phalloidin) selectively binds to F-actins.
  • Suitable for all fluorescent imaging platforms
  • Specific for actin in both living or fixed cells
  • Multiple applications
Caution: For research use only.