Human Recombinant FLT3 Protein (ECD, Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
10445-H02H1-50 10445-H02H1-50
103906-086EA 199.98 USD
103906-086 CA76479-746
Human Recombinant FLT3 Protein (ECD, Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human FLT3 (NP_004110.2) (Asn27-Ser543) was expressed with the Fc region of human IgG1 at the C-terminus.