Human Recombinant CD112/Nectin-2/PVRL2 Protein (from HEK293 Cells)
Supplier: Sino Biological
10005-HCCH-100 10005-HCCH-100
103620-682EA 300.98 USD
103620-682 CA103620-682
Human Recombinant CD112/Nectin-2/PVRL2 Protein (from HEK293 Cells)
Proteins and Peptides
The mature form of recombinant human CD112 (NP_002847.1) extracellular domain (Met 1-Leu 360) with quinary-aa peptide (DDDDK) at the C-terminus was expressed and purified.