Human Recombinant Layilin/LAYN Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
10208-H02H-100
103624-004EA 351.48 USD
103624-004
Human Recombinant Layilin/LAYN Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Glu 220) of human layilin (NP_849156.1) precursor was expressed with the C-terminal fused Fc region of human IgG1.