trFluor™ Eu-Streptavidin Conjugate (100 µg)
Supplier: AAT Bioquest
16925
76482-726EA 348.42 USD
76482-726
trFluor™ Eu-Streptavidin Conjugate (100 µg)
Proteins and Peptides
Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin.
  • Tr-fret reagent
  • Multiple applications
Caution: For research use only.