Casein, TAMRA-Conjugated (5 mg)
Supplier: AAT Bioquest
13441
76482-112EA 230.87 USD
76482-112
Casein, TAMRA-Conjugated (5 mg)
Proteins and Peptides
Casein is considered to be a generic substrate for a broad spectrum of proteases.
  • Excellent fluorescence intensity change
  • Signal directly proportional to protease activity
  • Bright signal
  • Ph-independent fluorescence
  • Suitable for low ph
Caution: For research use only.