Multi Vial Adapters, SPEX SamplePrep
Supplier: SPEX SamplePrep
8019 8018
76533-148EA 213.54 USD
76533-148 76533-150
Multi Vial Adapters, SPEX SamplePrep
Homogenizers Benchtop Homogenizers
Use adapters to run multiple samples or to hold vial securely in place during grinding.
  • Excellent for processing multiple smaller samples
Simultaneously run multiple vials in the Spex® SamplePrep high-energy Mixer/Mill® grinders.