APC-Cy7 Tandem fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

2625
76484-162EA 230.87 USD
76484-162
APC-Cy7 Tandem fluorescent probe
APC-Cy7 Tandem

Allophycocyanin (APC) is a phycobiliprotein isolated from Spirulina sp., a blue-green alga.


  • Suitable for most fluorescence platforms
  • Excellent signal
  • High quality

MW: 105000 g/mol
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/86/94/37378694.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR