PE-Cy7 Tandem fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

2616
76484-152EA 230.87 USD
76484-152
PE-Cy7 Tandem fluorescent probe
PE-Cy7 Tandem

PE-Cy7 is a popular color used in flow cytometry.


  • Suitable for most fluorescence platforms
  • Excellent signal
  • High quality

MW: 240000 g/mol
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/86/88/37378688.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR