APC-iFluor™ 750 Tandem, ReadiUse™ fluorescent probe, preactivated

Supplier: AAT BIOQUEST INC
Warning

2571
76481-598EA 405.91 USD
76481-598
APC-iFluor™ 750 Tandem, ReadiUse™ fluorescent probe, preactivated
APC-iFluor™ 750 Tandem

Allophycocyanin (APC) is a phycobiliprotein isolated from Spirulina sp., a blue-green alga.


  • Ready to use
  • High extinction coefficient
  • High quantum efficiency
  • Large Stokes shift
  • Spirulina

MW: 105000 g/mol
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/73/52/37377352.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR