Preactivated APC, ReadiUse™ fluorescent probe, preactivated
Supplier: AAT BIOQUEST INC
Warning
2561
76484-138EA 348.42 USD
76484-138
Preactivated APC, ReadiUse™ fluorescent probe, preactivated
Preactivated APC
Allophycocyanin (APC) is a phycobiliprotein isolated from Spirulina sp., a blue-green alga.
  • Ready to conjugate
  • No SMCC conjugation required
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/86/80/37378680.jpg