Cross linked-AlloPhycocyanin, ReadiUse™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

Synonyms: CL-APC

2503
76484-750EA 114.94 USD
76484-750
Cross linked-AlloPhycocyanin, ReadiUse™ fluorescent probe
Cross linked-AlloPhycocyanin

Cross linked-Allophycocyanin (CL-APC) is a phycobiliprotein isolated from Spirulina sp., a blue-green alga.


  • No dialysis required
  • Ready to use
  • High purity

MW: 105000 g/mol
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/89/51/37378951.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR