Tide Fluor™ 3WS succinimidyl ester, Tide Fluor™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

Synonyms: TF3WS SE

2346
76481-542EA 348.42 USD
76481-542
Tide Fluor™ 3WS succinimidyl ester, Tide Fluor™ fluorescent probe
Tide Fluor™ 3WS succinimidyl ester

Tide Fluor™ 3WS (TF3WS) family has the spectral properties essentially identical to those of Cy3.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

MW: 1024,31 g/mol
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/73/25/37377325.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR