Tide Fluor™ 8WS amine, Tide Fluor™ fluorescent probe
Supplier: AAT BIOQUEST INC
|
Warning
|
Synonyms:
TF8WS amine
2336
76481-532EA
230.87
USD
76481-532
Tide Fluor™ 8WS amine, Tide Fluor™ fluorescent probe
Tide Fluor™ 8WS amine
Tide Fluor™ 8WS (TF8WS) family has the spectral properties similar to those of IRDye 800.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity
MW:
1213,22 g/mol |
ADR:
5,II |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...