Tide Fluor™ 7WS maleimide, Tide Fluor™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

Synonyms: TF7WS maleimide

2332
76481-526EA 230.87 USD
76481-526
Tide Fluor™ 7WS maleimide, Tide Fluor™ fluorescent probe
Tide Fluor™ 7WS maleimide

Tide Fluor™ 7WS (TF7WS) family has the spectral properties similar to those of Cy7, IRDye 800 and Alexa Fluor 750.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

MW: 1013,08 g/mol
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/73/15/37377315.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR