Tide Fluor™ 8WS alkyne, Tide Fluor™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

Synonyms: TF8WS alkyne

2307
76481-520EA 230.87 USD
76481-520
Tide Fluor™ 8WS alkyne, Tide Fluor™ fluorescent probe
Tide Fluor™ 8WS alkyne

Tide Fluor™ 8WS (TF8WS) family has the spectral properties similar to those of IRDye 800.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

MW: 1094,18 g/mol
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/73/12/37377312.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR