Tide Fluor™ 3 phosphoramidite, Tide Fluor™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

Synonyms: TF3 CEP

2274
76484-774EA 464.68 USD
76484-774
Tide Fluor™ 3 phosphoramidite, Tide Fluor™ fluorescent probe
Tide Fluor™ 3 phosphoramidite

Tide Fluor™ 3 (TF3) phosphoramidite is a high-performance alternative to Cy3 phosphoramidite for the in-synthesis labeling of oligonucleotides.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

MW: 1045,36 g/mol
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/89/61/37378961.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR