Tide Fluor™ 1 maleimide, Tide Fluor™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

Synonyms: TF1 maleimide

2242
76481-450EA 230.87 USD
76481-450
Tide Fluor™ 1 maleimide, Tide Fluor™ fluorescent probe
Tide Fluor™ 1 maleimide

TF1 is designed to be a superior fluorophore alternative to EDANS.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

MW: 355,34 g/mol
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/72/72/37377272.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR