Tide Quencher™ 3WS acid, Tide Quencher™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

Synonyms: TQ3WS acid

2227
76481-426EA 230.87 USD
76481-426
Tide Quencher™ 3WS acid, Tide Quencher™ fluorescent probe
Tide Quencher™ 3WS acid

TQ3WS is designed to be a superior quencher to TAMRA, TF3 and Cy3.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

MW: 912 g/mol
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/72/60/37377260.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR