Tide Quencher™ 3 maleimide, Tide Quencher™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

Synonyms: TQ3 maleimide

2226
76481-424EA 230.87 USD
76481-424
Tide Quencher™ 3 maleimide, Tide Quencher™ fluorescent probe
Tide Quencher™ 3 maleimide

TQ3 is designed to be a superior quencher to TAMRA, TF3 and Cy3.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

MW: 575,64 g/mol
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/72/59/37377259.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR