Tide Quencher™ 3 CPG 1000 Å, Tide Quencher™ fluorescent probe
Supplier: AAT BIOQUEST INC
Warning
Synonyms: TQ3 CPG
2224
76481-422EA 206.72 USD
76481-422
Tide Quencher™ 3 CPG 1000 Å, Tide Quencher™ fluorescent probe
Tide Quencher™ 3 CPG
TQ3 is designed to be a superior quencher to TAMRA, TF3 and Cy3.
  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/72/58/37377258.jpg