Tide Quencher™ 2 phosphoramidite, Tide Quencher™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

Synonyms: TQ2 phosphoramidite

2208
76481-408EA 230.87 USD
76481-408
Tide Quencher™ 2 phosphoramidite, Tide Quencher™ fluorescent probe
Tide Quencher™ 2 phosphoramidite

TQ2 is designed to be a superior quencher to FAM, HEX, TET, JOE, TF2 and rhodamine 6G.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

MW: 681,87 g/mol
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/72/51/37377251.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR