Tide Quencher™ 1 azide, Tide Quencher™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

Synonyms: TQ1 azide

2188
76481-382EA 230.87 USD
76481-382
Tide Quencher™ 1 azide, Tide Quencher™ fluorescent probe
Tide Quencher™ 1 azide

TQ1 is designed to be a superior quencher alternative to DABCYL.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

MW: 358,42 g/mol
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/72/38/37377238.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR