Tide Quencher™ 7WS maleimide, Tide Quencher™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

Synonyms: TQ7WS maleimide

2109
76481-374EA 464.68 USD
76481-374
Tide Quencher™ 7WS maleimide, Tide Quencher™ fluorescent probe
Tide Quencher™ 7WS maleimide

TQ7WS is designed to be a superior quencher to Cy7, TF7 and Alexa Fluor 750.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

MW: 1109,24 g/mol
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/72/33/37377233.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR