Tide Quencher™ 6WS maleimide, Tide Quencher™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

Synonyms: TQ6WS maleimide

2094
76481-362EA 348.42 USD
76481-362
Tide Quencher™ 6WS maleimide, Tide Quencher™ fluorescent probe
Tide Quencher™ 6WS maleimide

TQ6WS is designed to be a superior quencher to Cy5.5, TF6 and Alexa Fluor 680.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

MW: 1132,24 g/mol
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/72/27/37377227.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR