Tide Quencher™ 4WS azide, Tide Quencher™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

Synonyms: TQ4WS azide

2068
76481-338EA 348.42 USD
76481-338
Tide Quencher™ 4WS azide, Tide Quencher™ fluorescent probe
Tide Quencher™ 4WS azide

TQ4WS is designed to be a superior quencher to ROX, TF4 and Texas Red®.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

MW: 968,24 g/mol
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/72/13/37377213.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR