iFluor® 488, Amine

Supplier: AAT BIOQUEST INC
Warning

1072
76481-188EA 230.87 USD
76481-188
iFluor® 488, Amine
iFluor®

Although FITC is still the most popular fluorescent labeling dye for preparing green fluorescent bioconjugates, there are certain limitations with FITC, such as severe photobleaching for microscope imaging and pH-sensitive fluorescence.


  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
  • pH independent fluorescence between 4-10

ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/71/34/37377134.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR