iFluor® 555, Maleimide

Supplier: AAT BIOQUEST INC
Warning

1063
76481-174EA 230.87 USD
76481-174
iFluor® 555, Maleimide
iFluor®

AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.


  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
  • pH independent fluorescence between 3 to 11

ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/71/28/37377128.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR