Staurosporine 1 mM, ReadiUse™ for apoptosis positive control
Supplier: AAT BIOQUEST INC
|
Warning
|
Synonyms:
Antibiotic AM-2282
80050
76484-814EA
114.94
USD
76484-814
Staurosporine 1 mM, ReadiUse™ for apoptosis positive control
Staurosporine
Staurosporine is widely used as a positive control for inducing apoptosis.
- Well-cited positive apoptosis control
- Simple effective apoptosis induction in multiple cell lines
- Cell permeable and reversible
- Inhibits some types of platelet aggregation
Formula:
C₂₈H₂₆N₄O₃ MW: 466.541 g/mol |
MDL Number:
MFCD18252446 CAS Number: 62996-74-1 ADR: 5,II |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...