Acridinium C2 NHS Ester for biomolecular labeling
Supplier: AAT BIOQUEST INC
|
Warning
|
Synonyms:
Acridinium C2 N-hydroxysuccinimide ester
, 4-(2-Succinimidyl-oxycarbonylethyl)-phenyl-10-acridinium-9-carboxylate Triflurormethyl Sulfonate
26015
76484-996EA
230.87
USD
76484-996
Acridinium C2 NHS Ester for biomolecular labeling
Acridinium C2 NHS Ester
The acridinium NHS ester can be used to label proteins and nucleic acids.
- Amine reactive
- Luminescent detection
- Sensitive detection
Formula:
C29H23F3N2O9S MW: 632.56 g/mol |
CAS Number:
177332-37-5 ADR: 5,II |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...