DiSBAC2(3) for membrane potential measurement

Supplier: AAT BIOQUEST INC
Warning

Synonyms: Bis-(1,3-diethylthiobarbituric acid)trimethine oxonol

21414
76483-388EA 114.94 USD
76483-388
DiSBAC2(3) for membrane potential measurement
DiSBAC2(3)

DiSBAC2(3) is a sensitive slow-response membrane potential probe that is widely used for measuring membrane potentials of many biological systems.


  • Sensitive detection
  • Slow response probe
  • Multiple applications

Formula: C₁₉H₂₄N₄O₄S₂
MW: 436,55 g/mol
CAS Number: 47623-98-3
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/82/18/37378218.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR