DiBAC₄(3) for membrane potential measurement

Supplier: AAT BIOQUEST INC
Warning

Synonyms: Bis(1,3-dibutylbarbituric acid) trimethine oxonolDiBAC4(3)

21411
76483-386EA 114.94 USD
76483-386
DiBAC₄(3) for membrane potential measurement
DiBAC₄(3)

DiBAC4(3) is a sensitive slow-response probe for measuring cellular membrane potential.


  • Sensitive detection
  • Slow response probe
  • Multiple applications

Formula: C₂₇H₄₀N₄O₆
MW: 516.63 g/mol
MDL Number: MFCD07437857
CAS Number: 70363-83-6
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/82/14/37378214.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR