Mca-PLAQAV-Dpa-RSSSR-NH2
Supplier: AAT BIOQUEST INC
Warning
13560
76485-626EA 163.8 USD
76485-626
Mca-PLAQAV-Dpa-RSSSR-NH2
Mca-PLAQAV-Dpa-RSSSR-NH2
Mca-PLAQAV-Dpa-RSSSR-NH2 is a fluorogenic peptide substrate for detecting tumor necrosis factor-alpha converting enzyme (TACE or ADAM17) and related enzymes such as ADAM8, ADAM9 and ADAM10.
  • tumor necrosis factor-alpha converting enzyme detection
  • bright signal
ADR: 5,II