(Ac-ANW)2R110
Supplier: AAT BIOQUEST INC
Warning
13455
76482-122EA 230.87 USD
76482-122
(Ac-ANW)2R110
(Ac-ANW)2R110
(Ac-ANW)2R110 is a selective fluorogenic substrate for proteasome 20S-beta 5i.
  • highly sensitive substrate
  • bright signal
  • multiple applications
ADR: 5,II