Z-IETD-FMK
Supplier: AAT BIOQUEST INC
Warning
13305
76485-300EA 107.15 USD
76485-300
Z-IETD-FMK
Z-IETD-FMK
Z-IETD-FMK is a cell-permeable, irreversible caspase-8 inhibitor.
  • caspase inhibitor
  • enhanced stability
  • increased cell permeability
ADR: 5,II