Beta-Ala-R110
Supplier: AAT BIOQUEST INC
Warning
13203
76485-688EA 219.43 USD
76485-688
Beta-Ala-R110
Beta-Ala-R110
Beta-Alanine-R110 is a fluorogenic substrate that might be used for detection pancreatic elastase, arylamidase and beta-alanine aminopeptidase activities.
  • adaptable fluorogenic substrate
  • detects enzymes in multiple species
  • suitable for cell, food, and other sample types
ADR: 5,II