BOC-Val-Pro-Arg-Aminoluciferin
Supplier: AAT BIOQUEST INC
Warning
13002
76482-052EA 580.89 USD
76482-052
BOC-Val-Pro-Arg-Aminoluciferin
BOC-Val-Pro-Arg-Aminoluciferin
BOC-Val-Pro-Arg-Aminoluciferin is a sentive luminescent substrate for alpha-thrombin.
  • high sensitivity
  • excellent quality luminogenic substrate
  • adaptable to multiple types of luciferase assays
ADR: 5,II