Cy3 Bis NTA-Ni complex, HIS Lite™
Supplier: AAT BIOQUEST INC
Warning
12610
76482-044EA 464.68 USD
76482-044
Cy3 Bis NTA-Ni complex, HIS Lite™
Cy3 Bis NTA-Ni
Polyhistidine is one of the most popular affinity tags incorporated into recombinant proteins.
  • useful for detecting histidine-tagged proteins
  • well-cited building block
  • multiple applications
ADR: 5,II