Rhodamine aldehyde [5-TAMRA aldehyde] for biomolecular labeling
Supplier: AAT BIOQUEST INC
Warning
9005
76485-706EA 331.78 USD
76485-706
Rhodamine aldehyde [5-TAMRA aldehyde] for biomolecular labeling
Rhodamine aldehyde [5-TAMRA aldehyde]
Rhodamine aldehyde is a reactive fluorescent dye that can react with an amine, hydrazine or hydroxylamine. Aldehyde group is reactive toward amines, hydrazide or hydroxylamine groups from pH 5-9. Unlike amine reactive succinimidyl ester group (NHS), aldehyde can react with N-terminal amine groups at acidic pH, a condition sometimes required for certain bioconjugation reactions. Aldehyde reacts with amine group to form an intermediate Schiff bond. Further reduction with hydride will form a stable C-N bond. Reaction between aldehyde and other groups allows site-specific conjugation and labeling of fluorescein dyes to desired position on targeted molecules. Conjugated rhodamine dye can be easily detected by a common fluorescence instrument under TRITC or Cy3 channel.
  • Reactive with amine, hydrazine, or hydroxylamine
  • Detectable with the FITC channel
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/95/02/37379502.jpg