CDPI3 Maleimide DNA binding
Supplier: AAT BIOQUEST INC
Warning
6908
76485-518EA 219.43 USD
76485-518
CDPI3 Maleimide DNA binding
CDPI3 Maleimide
Minor Groove Binder (MGB) molecules selectively bind to the minor groove of a DNA molecule, the shallow furrow in the DNA helix.
  • Stable binding
  • Hybridization probe precursor
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/94/04/37379404.jpg