CDPI3 Succinimidyl Ester DNA binding
Supplier: AAT BIOQUEST INC
Warning
6907
76485-516EA 219.43 USD
76485-516
CDPI3 Succinimidyl Ester DNA binding
CDPI3 Succinimidyl Ester
Minor Groove Binder (MGB) molecules selectively bind to the minor groove of a DNA molecule, the shallow furrow in the DNA helix.
  • Stable binding
  • Hybridization probe precursor
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/94/03/37379403.jpg