6-TAMRA CPG 1000 Å for oligonucleotide labeling
Supplier: AAT BIOQUEST INC
Warning
6051
76481-842EA 580.89 USD
76481-842
6-TAMRA CPG 1000 Å for oligonucleotide labeling
6-TAMRA CPG
The light-absorbing properties of TAMRA, and spectral overlap with several commonly used fluorophores - including FAM, HEX, TET and JOE, make it useful as a FRET acceptor for the dual labeled FRET probes such as Molecular Beacons.
  • FRET building block
  • Multiple applications
ADR: 5,II
https://us.vwr.com/stibo/web/std.lang.all/74/75/37377475.jpg